3,7-Dibromo-5,5-dioctyl-5H-dibenzo[b,d]silole 95%

BD254692-250MG BD254692-100MG BD254692-1G
BLDPBD254692-250MGEA 147 EUR
BLDPBD254692-250MG BLDPBD254692-100MG BLDPBD254692-1G
3,7-Dibromo-5,5-dioctyl-5H-dibenzo[b,d]silole 95%
3,7-Dibromo-5,5-dioctyl-5H-dibenzo[b,d]silole

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₈H₄₀Br₂Si
MW: 564,51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619361
CAS Number: 891182-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR