(R)-1-(3-Methoxyphenyl)ethanamine hydrochloride 95%

BD254988-5G BD254988-1G BD254988-250MG
BLDPBD254988-5GEA 432 EUR
BLDPBD254988-5G BLDPBD254988-1G BLDPBD254988-250MG
(R)-1-(3-Methoxyphenyl)ethanamine hydrochloride 95%
(R)-1-(3-Methoxyphenyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₁₄ClNO
MW: 187,66 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11101207
CAS Number: 1167414-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR