(S)-1-(3-Fluorophenyl)propan-1-amine hydrochloride 95%

BD255642-250MG BD255642-100MG
BLDPBD255642-250MGEA 205 EUR
BLDPBD255642-250MG BLDPBD255642-100MG
(S)-1-(3-Fluorophenyl)propan-1-amine hydrochloride 95%
(S)-1-(3-Fluorophenyl)propan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₃ClFN
MW: 189,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12757046
CAS Number: 1310923-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR