7-Hydroxy-5-methoxy-2,3-dihydro-1H-inden-1-one 95%

BLDPBD255827-1GEA 394 EUR
BLDPBD255827-1G
7-Hydroxy-5-methoxy-2,3-dihydro-1H-inden-1-one 95%
7-Hydroxy-5-methoxy-2,3-dihydro-1H-inden-1-one

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₀O₃
MW: 178,18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18208821
CAS Number: 76842-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR