5-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazin-2-amine 95%

BD256371-100MG BD256371-250MG
BLDPBD256371-100MGEA 152 EUR
BLDPBD256371-100MG BLDPBD256371-250MG
5-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazin-2-amine 95%
5-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazin-2-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₁₂N₄
MW: 152,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17215187
CAS Number: 1227210-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR