3-(Hydroxymethyl)-5-methylphenol 95+%

BD256400-100MG BD256400-50MG
BLDPBD256400-100MGEA 131 EUR
BLDPBD256400-100MG BLDPBD256400-50MG
3-(Hydroxymethyl)-5-methylphenol 95+%
3-(Hydroxymethyl)-5-methylphenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₀O₂
MW: 138,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397252
CAS Number: 606488-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR