(2S,3R,5S)-5-(4-Amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol 95+%

BLDPBD256630-1GEA 75.6 EUR
BLDPBD256630-1G BLDPBD256630-250MG BLDPBD256630-100MG
(2S,3R,5S)-5-(4-Amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol 95+%
(2S,3R,5S)-5-(4-Amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₁H₁₃IN₄O₃
MW: 376,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07778650
CAS Number: 908130-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR