Methyl-2,3-dimethylbenzoate 97%

Synonyms: Methyl 2,3-dimethylbenzoate

BLDPBD257933-10GEA 76.7 EUR
BLDPBD257933-10G BLDPBD257933-100G BLDPBD257933-5G BLDPBD257933-1G BLDPBD257933-25G
Methyl-2,3-dimethylbenzoate 97%
Methyl 2,3-dimethylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₂O₂
MW: 164,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203773
CAS Number: 15012-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR