Methyl 3-((tert-butoxycarbonyl)amino)thiophene-2-carboxylate 95%

BD259219-5G BD259219-1G BD259219-250MG
BLDPBD259219-5GEA 124 EUR
BLDPBD259219-5G BLDPBD259219-1G BLDPBD259219-250MG
Methyl 3-((tert-butoxycarbonyl)amino)thiophene-2-carboxylate 95%
Methyl 3-((tert-butoxycarbonyl)amino)thiophene-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₁H₁₅NO₄S
MW: 257,31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01763684
CAS Number: 149587-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR