Ethyl 7-bromoquinoline-3-carboxylate 98%

BD259587-5G BD259587-250MG BD259587-1G
BLDPBD259587-5GEA 937 EUR
BLDPBD259587-5G BLDPBD259587-250MG BLDPBD259587-1G
Ethyl 7-bromoquinoline-3-carboxylate 98%
Ethyl 7-bromoquinoline-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₀BrNO₂
MW: 280,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16657753
CAS Number: 1226762-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR