1-Chloro-3-methylbutan-2-one 95%

BLDPBD259785-1GEA 533 EUR
BLDPBD259785-1G BLDPBD259785-100MG BLDPBD259785-250MG
1-Chloro-3-methylbutan-2-one 95%
1-Chloro-3-methylbutan-2-one

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₉ClO
MW: 120,57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19160714
CAS Number: 17687-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR