3-Fluoro-4-thiomorpholinoaniline 95%

BD260643-1G BD260643-25G BD260643-5G
BLDPBD260643-1GEA 27.3 EUR
BLDPBD260643-1G BLDPBD260643-25G BLDPBD260643-5G
3-Fluoro-4-thiomorpholinoaniline 95%
3-Fluoro-4-thiomorpholinoaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₃FN₂S
MW: 212,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953533
CAS Number: 237432-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR