4-Amino-3-chloro-5-fluorobenzamide 95%

BLDPBD260684-1GEA 307 EUR
BLDPBD260684-1G BLDPBD260684-250MG BLDPBD260684-100MG
4-Amino-3-chloro-5-fluorobenzamide 95%
4-Amino-3-chloro-5-fluorobenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₆ClFN₂O
MW: 188,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27922987
CAS Number: 1823346-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR