2-Bromo-1-(1-methyl-1H-pyrrol-2-yl)ethanone 95%

BD260787-100MG
BLDPBD260787-100MGEA 80.9 EUR
BLDPBD260787-100MG
2-Bromo-1-(1-methyl-1H-pyrrol-2-yl)ethanone 95%
2-Bromo-1-(1-methyl-1H-pyrrol-2-yl)ethanone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₈BrNO
MW: 202,04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12797405
CAS Number: 65438-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR