4-Bromo-1H-pyrrolo[2,3-b]pyridin-3-amine 95%

BD260895-100MG BD260895-250MG BD260895-1G
BLDPBD260895-100MGEA 123 EUR
BLDPBD260895-100MG BLDPBD260895-250MG BLDPBD260895-1G
4-Bromo-1H-pyrrolo[2,3-b]pyridin-3-amine 95%
4-Bromo-1H-pyrrolo[2,3-b]pyridin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₆BrN₃
MW: 212,04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09880125
CAS Number: 943323-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR