N-Cyclohexyl-3-boronobenzenesulphonamide 97%

Synonyms: (3-(N-Cyclohexylsulphamoyl)phenyl)boronic acid

BLDPBD260898-100MGEA 41 EUR
BLDPBD260898-100MG BLDPBD260898-250MG
N-Cyclohexyl-3-boronobenzenesulphonamide 97%
N-Cyclohexyl 3-boronobenzenesulphonamide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₈BNO₄S
MW: 283,1516 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07783859
CAS Number: 871329-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR