3-(2-Chlorobenzyloxy)phenylboronic acid 98%

Synonyms: (3-((2-Chlorobenzyl)oxy)phenyl)boronic acid

BD261016-250MG BD261016-1G BD261016-5G
BLDPBD261016-250MGEA 27.3 EUR
BLDPBD261016-250MG BLDPBD261016-1G BLDPBD261016-5G
3-(2-Chlorobenzyloxy)phenylboronic acid 98%
3-(2-Chlorobenzyloxy)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₁₂BClO₃
MW: 262,5000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05865198
CAS Number: 845551-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR