Methyl 2-methyl-2-(2-nitrophenyl)propanoate 97%

BLDPBD261125-100MGEA 36.8 EUR
BLDPBD261125-100MG BLDPBD261125-1G BLDPBD261125-250MG
Methyl 2-methyl-2-(2-nitrophenyl)propanoate 97%
Methyl 2-methyl-2-(2-nitrophenyl)propanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₃NO₄
MW: 223,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16620966
CAS Number: 136764-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR