3-Iodo-5-methyl-1H-pyrrolo[3,2-b]pyridine 95%

BLDPBD261319-100MGEA 125 EUR
BLDPBD261319-100MG BLDPBD261319-250MG BLDPBD261319-1G
3-Iodo-5-methyl-1H-pyrrolo[3,2-b]pyridine 95%
3-Iodo-5-methyl-1H-pyrrolo[3,2-b]pyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₈H₇IN₂
MW: 258,06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09880091
CAS Number: 1000343-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR