4-((4-Methoxyphenyl)amino)benzonitrile 97%

BD261630-100MG BD261630-250MG BD261630-1G
BLDPBD261630-100MGEA 200 EUR
BLDPBD261630-100MG BLDPBD261630-250MG BLDPBD261630-1G
4-((4-Methoxyphenyl)amino)benzonitrile 97%
4-((4-Methoxyphenyl)amino)benzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₄H₁₂N₂O
MW: 224,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11135494
CAS Number: 319016-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR