2-Ethynyl-3-methoxypyridine 97%

BLDPBD262121-250MGEA 173 EUR
BLDPBD262121-250MG BLDPBD262121-1G BLDPBD262121-100MG
2-Ethynyl-3-methoxypyridine 97%
2-Ethynyl-3-methoxypyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₇NO
MW: 133,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18256372
CAS Number: 1256805-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR