(E)-4-((4-Hydroxyphenyl)diazenyl)benzoic acid 96%
|
|
BLDPBD262142-1GEA
191
EUR
BLDPBD262142-1G
BLDPBD262142-100MG
BLDPBD262142-250MG
(E)-4-((4-Hydroxyphenyl)diazenyl)benzoic acid 96%
(E)-4-((4-Hydroxyphenyl)diazenyl)benzoic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₃H₁₀N₂O₃ MW: 242,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12198372 CAS Number: 105299-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...