(S)-3-Hydroxy-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide 98%

BD262311-50MG
BLDPBD262311-50MGEA 624 EUR
BLDPBD262311-50MG
(S)-3-Hydroxy-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide 98%
(S)-3-Hydroxy-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₅H₂₂N₂O₂
MW: 382,45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00944075
CAS Number: 174636-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR