5-Chloro-3-phenylisoxazole 97%

BD262489-100MG BD262489-250MG
BLDPBD262489-100MGEA 152 EUR
BLDPBD262489-100MG BLDPBD262489-250MG
5-Chloro-3-phenylisoxazole 97%
5-Chloro-3-phenylisoxazole

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆ClNO
MW: 179,60 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00046079
CAS Number: 3356-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR