(2S,3R,4S,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol 95%

BLDPBD262819-250MGEA 27.3 EUR
BLDPBD262819-250MG BLDPBD262819-5G BLDPBD262819-1G BLDPBD262819-25G BLDPBD262819-10G BLDPBD262819-100G
(2S,3R,4S,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol 95%
(2S,3R,4S,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₂H₂₇ClO₇
MW: 438,89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27923102
CAS Number: 714269-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR