6-Chloro-2-phenylpyridazin-3(2H)-one 95+%

BD263010-250MG BD263010-1G BD263010-100MG
BLDPBD263010-250MGEA 184 EUR
BLDPBD263010-250MG BLDPBD263010-1G BLDPBD263010-100MG
6-Chloro-2-phenylpyridazin-3(2H)-one 95+%
6-Chloro-2-phenylpyridazin-3(2H)-one

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₇ClN₂O
MW: 206,62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00462958
CAS Number: 1698-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR