Methyl 2-bromo-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate 95%

BD263889-250MG BD263889-100MG BD263889-1G
BLDPBD263889-250MGEA 42 EUR
BLDPBD263889-250MG BLDPBD263889-100MG BLDPBD263889-1G
Methyl 2-bromo-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate 95%
Methyl 2-bromo-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₈BrNO₄S
MW: 294,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195322
CAS Number: 365996-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR