5-Bromo-4-phenylthiazol-2-amine 95%

BD264223-250MG BD264223-100MG BD264223-1G
BLDPBD264223-250MGEA 110 EUR
BLDPBD264223-250MG BLDPBD264223-100MG BLDPBD264223-1G
5-Bromo-4-phenylthiazol-2-amine 95%
5-Bromo-4-phenylthiazol-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₇BrN₂S
MW: 255,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07364351
CAS Number: 61954-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR