1H-Pyrrol-3-amine hydrochloride 95%

BD264311-250MG
BLDPBD264311-250MGEA 460 EUR
BLDPBD264311-250MG
1H-Pyrrol-3-amine hydrochloride 95%
1H-Pyrrol-3-amine hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₄H₇ClN₂
MW: 118,56 g/mol
Storage Temperature: Freezer
MDL Number: MFCD27920766
CAS Number: 1956319-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR