Methyl 6-acetylnicotinate 95%

BLDPBD264628-250MGEA 134 EUR
BLDPBD264628-250MG BLDPBD264628-100MG BLDPBD264628-1G
Methyl 6-acetylnicotinate 95%
Methyl 6-acetylnicotinate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉NO₃
MW: 179,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036565
CAS Number: 153559-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR