1-Bromo-2-chloro-3,5-dimethylbenzene 97%

BD265494-1G
BLDPBD265494-1GEA 131 EUR
BLDPBD265494-1G
1-Bromo-2-chloro-3,5-dimethylbenzene 97%
1-Bromo-2-chloro-3,5-dimethylbenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrCl
MW: 219,51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780645
CAS Number: 933585-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR