Methyl 2,2-bis((methoxycarbonyl)amino)acetate 95%

BLDPBD265703-1GEA 364 EUR
BLDPBD265703-1G BLDPBD265703-250MG BLDPBD265703-5G
Methyl 2,2-bis((methoxycarbonyl)amino)acetate 95%
Methyl 2,2-bis((methoxycarbonyl)amino)acetate

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₂N₂O₆
MW: 220,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699145
CAS Number: 60794-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR