Phenyl(pyridin-4-yl)methanamine dihydrochloride 95%

BLDPBD265803-250MGEA 131 EUR
BLDPBD265803-250MG BLDPBD265803-1G BLDPBD265803-100MG
Phenyl(pyridin-4-yl)methanamine dihydrochloride 95%
Phenyl(pyridin-4-yl)methanamine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₁₄Cl₂N₂
MW: 257,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00763280
CAS Number: 423761-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR