1-(6-Aminonaphthalen-2-yl)ethanone 95%

BD265878-1G BD265878-100MG BD265878-250MG
BLDPBD265878-1GEA 565 EUR
BLDPBD265878-1G BLDPBD265878-100MG BLDPBD265878-250MG
1-(6-Aminonaphthalen-2-yl)ethanone 95%
1-(6-Aminonaphthalen-2-yl)ethanone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₂H₁₁NO
MW: 185,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20693622
CAS Number: 7470-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR