Ethyl 4-formylpicolinate 95%

BLDPBD266663-100MGEA 111 EUR
BLDPBD266663-100MG BLDPBD266663-250MG BLDPBD266663-1G
Ethyl 4-formylpicolinate 95%
Ethyl 4-formylpicolinate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₉NO₃
MW: 179,17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19690341
CAS Number: 189819-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR