(5,6-Dichloropyridin-2-yl)methanamine 95%

BLDPBD266678-250MGEA 0 EUR
BLDPBD266678-250MG BLDPBD266678-100MG
(5,6-Dichloropyridin-2-yl)methanamine 95%
(5,6-Dichloropyridin-2-yl)methanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₆Cl₂N₂
MW: 177,03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18254247
CAS Number: 1256825-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR