2-(Boc-aminomethyl)benzyl Alcohol 98%

BD266795-1G BD266795-100MG BD266795-250MG
BLDPBD266795-1GEA 86.1 EUR
BLDPBD266795-1G BLDPBD266795-100MG BLDPBD266795-250MG
2-(Boc-aminomethyl)benzyl Alcohol 98%
2-(Boc-aminomethyl)benzyl Alcohol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₉NO₃
MW: 237,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22418063
CAS Number: 1333114-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR