4'-Bromo-[1,1'-biphenyl]-2,4-diol 95%

BD268671-250MG BD268671-100MG BD268671-5G BD268671-1G
BLDPBD268671-250MGEA 49.4 EUR
BLDPBD268671-250MG BLDPBD268671-100MG BLDPBD268671-5G BLDPBD268671-1G
4'-Bromo-[1,1'-biphenyl]-2,4-diol 95%
4'-Bromo-[1,1'-biphenyl]-2,4-diol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₉BrO₂
MW: 265,10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23381094
CAS Number: 1418117-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR