5-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one 97%

BLDPBD269974-5GEA 737 EUR
BLDPBD269974-5G BLDPBD269974-100MG BLDPBD269974-250MG BLDPBD269974-1G
5-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one 97%
5-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₇BrN₂O
MW: 227,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00504782
CAS Number: 84712-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR