5-Chloro-3-methyl-2,3-dihydro-1H-inden-1-one 95+%
|
|
BLDPBD270519-100MGEA
267
EUR
BLDPBD270519-100MG
BLDPBD270519-1G
BLDPBD270519-250MG
5-Chloro-3-methyl-2,3-dihydro-1H-inden-1-one 95+%
5-Chloro-3-methyl-2,3-dihydro-1H-inden-1-one
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₉ClO MW: 180,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27996953 CAS Number: 66248-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...