6-Methoxychroman-2-one 95%

BLDPBD271276-100MGEA 89.3 EUR
BLDPBD271276-100MG BLDPBD271276-1G BLDPBD271276-250MG
6-Methoxychroman-2-one 95%
6-Methoxychroman-2-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀O₃
MW: 178,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28016649
CAS Number: 20920-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR