4-Fluoropyrazolo[1,5-a]pyridine-3-carboxylic acid 98%

BD272073-250MG BD272073-1G BD272073-100MG
BLDPBD272073-250MGEA 257 EUR
BLDPBD272073-250MG BLDPBD272073-1G BLDPBD272073-100MG
4-Fluoropyrazolo[1,5-a]pyridine-3-carboxylic acid 98%
4-Fluoropyrazolo[1,5-a]pyridine-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₅FN₂O₂
MW: 180,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24481556
CAS Number: 1352625-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR