N-[3-(Trifluoromethyl)benzyl]ethylamine 97%

BD273288-1G BD273288-5G BD273288-10G BD273288-250MG BD273288-25G
BLDPBD273288-1GEA 60.9 EUR
BLDPBD273288-1G BLDPBD273288-5G BLDPBD273288-10G BLDPBD273288-250MG BLDPBD273288-25G
N-[3-(Trifluoromethyl)benzyl]ethylamine 97%
N-[3-(Trifluoromethyl)benzyl]ethylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₂F₃N
MW: 203,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09935363
CAS Number: 14355-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR