6-(2-Furyl)picolinaldehyde 98%

Synonyms: 6-(Furan-2-yl)picolinaldehyde

BLDPBD273443-250MGEA 117 EUR
BLDPBD273443-250MG BLDPBD273443-1G BLDPBD273443-100MG
6-(2-Furyl)picolinaldehyde 98%
6-(2-Furyl)picolinaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₇NO₂
MW: 173,168 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06410008
CAS Number: 208110-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR