1-(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine 95%

BD273705-1G BD273705-100MG BD273705-250MG
BLDPBD273705-1GEA 185 EUR
BLDPBD273705-1G BLDPBD273705-100MG BLDPBD273705-250MG
1-(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine 95%
1-(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₇H₂₅BClNO₂
MW: 321,64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18087735
CAS Number: 1256360-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR