(9,9-Didodecyl-9H-fluorene-2,7-diyl)diboronic acid 95%

BLDPBD273729-250MGEA 55.7 EUR
BLDPBD273729-250MG BLDPBD273729-100MG BLDPBD273729-1G
(9,9-Didodecyl-9H-fluorene-2,7-diyl)diboronic acid 95%
(9,9-Didodecyl-9H-fluorene-2,7-diyl)diboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₃₇H₆₀B₂O₄
MW: 590,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701624
CAS Number: 480424-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR