N-[1-(Hydroxymethyl)propyl]-4-methylbenzenesulphonamide 95%

BLDPBD273796-100MGEA 23.1 EUR
BLDPBD273796-100MG BLDPBD273796-250MG
N-[1-(Hydroxymethyl)propyl]-4-methylbenzenesulphonamide 95%
N-[1-(Hydroxymethyl)propyl]-4-methylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₇NO₃S
MW: 243,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00458993
CAS Number: 304460-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR