(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid 98%

BLDPBD274006-100MGEA 42 EUR
BLDPBD274006-100MG BLDPBD274006-5G BLDPBD274006-250MG BLDPBD274006-1G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₀H₂₁NO₅
MW: 355,38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270543
CAS Number: 173212-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR