2-(2-Bromo-4-methoxyphenoxy)acetonitrile 98%

BD275924-1G BD275924-5G BD275924-250MG
BLDPBD275924-1GEA 55.7 EUR
BLDPBD275924-1G BLDPBD275924-5G BLDPBD275924-250MG
2-(2-Bromo-4-methoxyphenoxy)acetonitrile 98%
2-(2-Bromo-4-methoxyphenoxy)acetonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈BrNO₂
MW: 242,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09906448
CAS Number: 951918-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR