3-Cyanobenzamide 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD279452-5GEA 340 EUR
BLDPBD279452-5G BLDPBD279452-250MG BLDPBD279452-1G
3-Cyanobenzamide 97%
3-Cyanobenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆N₂O
MW: 146,1460 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017574
CAS Number: 3441-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR